Learn More
Aviva Systems Biology Corp CCDC117 Antibody - middle region (ARP53255_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53255_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC117. It was validated on Western Blot. . Alternate names: FLJ33814, dJ366L4.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-285-816