Learn More
Aviva Systems Biology Corp CCDC127 Antibody - N-terminal region (ARP52839_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52839_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC127. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CCDC127. Alternate names: FLJ25701ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-773