Learn More
Aviva Systems Biology Corp CCDC146 Antibody - middle region (ARP57476_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57476_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC146. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CCDC146. Alternate names: KIAA1505ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-286-318