Learn More
Aviva Systems Biology Corp CCDC16 Antibody - middle region (ARP36112_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36112_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCDC16. Alternate names: OMCG1, SEL13, CCDC16ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-56018