Learn More
Aviva Systems Biology Corp CFAP45 Antibody - N-terminal region (ARP51321_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51321_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC19. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CCDC19. Alternate names: NESG1, RP11-190A12.6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rat, Yeast
Catalog No. 50-014-60545