Learn More
Aviva Systems Biology Corp CCDC38 Antibody - N-terminal region (ARP53418_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53418_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC38. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CCDC38. Alternate names: FLJ40089ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61021