Learn More
Aviva Systems Biology Corp CCDC43 Antibody - N-terminal region (ARP51897_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51897_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC43. It was validated on Western Blot. . Alternate names: FLJ31795ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-285-605