Learn More
Aviva Systems Biology Corp CCDC46 Antibody - C-terminal region (ARP53824_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53824_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC46. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CCDC46. Alternate names: FLJ39610, MGC33887, CCDC46, MACOCOApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61139