Learn More
Aviva Systems Biology Corp CCDC52 Antibody - middle region (ARP52757_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52757_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC52. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCDC52. Alternate names: FLJ26064, FLJ44949, SPICE, CCDC52ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60823