Learn More
Aviva Systems Biology Corp CCDC54 Antibody - N-terminal region (ARP58605_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58605_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC54. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CCDC54. Alternate names: FLJ25362, NYD-SP17ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62372