Learn More
Aviva Systems Biology Corp CCDC63 Antibody - middle region (ARP53051_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53051_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC63. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCDC63. Alternate names: FLJ35843ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60905