Learn More
Aviva Systems Biology Corp CCDC74A Antibody - middle region (ARP58438_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58438_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC74A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCDC74A. Alternate names: FLJ40345, CCDC74AV2, CCDC74AV3ApplicationWB. Species reactivity: Horse, Human, Mouse, Rabbit
Catalog No. 50-014-62308