Learn More
Aviva Systems Biology Corp CCDC76 Antibody - N-terminal region (ARP57327_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57327_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC76. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CCDC76. Alternate names: FLJ10287, FLJ11219, CCDC76ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-299