Learn More
Aviva Systems Biology Corp CCDC78 Antibody - middle region (ARP53233_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53233_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC78. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCDC78. Alternate names: C16orf25, FLJ34512, JFP10, LA16c-444G9.2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60981