Learn More
Aviva Systems Biology Corp CCDC85B Antibody - N-terminal region (ARP62259_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62259_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC85B. It was validated on Western Blot. . Alternate names: DIPAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Yeast, Zebrafish
Catalog No. 50-287-633