Learn More
Aviva Systems Biology Corp CCDC87 Antibody - C-terminal region (ARP57158_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57158_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC87. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CCDC87. Alternate names: FLJ10786ApplicationWB. Species reactivity: Horse, Human, Pig
Catalog No. 50-286-269