Learn More
Aviva Systems Biology Corp CCDC90A Antibody - middle region (ARP44777_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44777_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC90A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCDC90A. Alternate names: C6orf79, FLJ20958, CCDC90AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-58893