Learn More
Aviva Systems Biology Corp Ccdc91 Antibody - middle region (ARP57198_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57198_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ccdc91. It was validated on Western Blot. . Alternate names: 1700086G08Rik, 1810060J02Rik, AU045683, AW555238ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-275