Learn More
Biolog Inc CCFA-HT - Cycloserine Cefoxitin Fructose Agar with Horse Blood and Taruocholate, 1 Plate/Pkg, Refrigerated must ship overnight
SDP
Catalog No. NC0097254
Encompass
This item has a minimum qty of 8 per supplier requirements.
Cycloserine-Cefoxitin Fructose Agar w/ Horse Blood & Taurocholate (CCFA-HT) is an enriched selective media used for the isolation & presumptive identification of C. difficile. This media is prepared, dispensed, & packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.CCFA-HT is an enriched selective media used for the isolation of C. difficile. The nutritive base consists of animal peptones & fructose & is supplemented w/ cefoxitin & cycloserine at concentrations that inhibit the growth of most normal fecal flora. Cycloserine will inhibit gram-negative bacteria, while cefoxitin will inhibit both gram-positive & gram-negative organisms. Horse blood is added to stimulate growth. Sodium Taurocholate is added to help in the germination of spores.
Catalog No. NC0097254 Supplier Biolog Inc Supplier No. AS2136
This item has a minimum qty of 8 per supplier requirements.
Cycloserine-Cefoxitin Fructose Agar w/ Horse Blood & Taurocholate (CCFA-HT) is an enriched selective media used for the isolation & presumptive identification of C. difficile. This media is prepared, dispensed, & packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.CCFA-HT is an enriched selective media used for the isolation of C. difficile. The nutritive base consists of animal peptones & fructose & is supplemented w/ cefoxitin & cycloserine at concentrations that inhibit the growth of most normal fecal flora. Cycloserine will inhibit gram-negative bacteria, while cefoxitin will inhibit both gram-positive & gram-negative organisms. Horse blood is added to stimulate growth. Sodium Taurocholate is added to help in the germination of spores.