Learn More
Selleck Chemical LLC CCG-1423-10mg
SDP
Catalog No. 502389593
Encompass_Preferred
CCG-1423 is a specific RhoA pathway inhibitor, which inhibits SRF-mediated transcription.
Catalog No. 50-238-9593 Supplier Selleck Chemical LLC Supplier No. S771910MG
CCG-1423 is a specific RhoA pathway inhibitor, which inhibits SRF-mediated transcription.