Learn More
Enzo Life Sciences CCG-1423 (10mg). CAS: 285986-88-1
SDP
Supplier:  Enzo Life Sciences BMLEI3940010
Encompass_Preferred
Cell permeable inhibitor of RhoA- and RhoC-mediated signaling. Inhibits LPA-induced DNA synthesis in PC-3 prostate cancer cell lines (<1µM) and inhibits the growth of RhoC-overexpressing melanoma cell lines (A375M2 and SK-Mel-147) at nanomolar concentrations. Alternative name: N-(2-(4-Chloroanilino)-1-methyl-2-oxoethoxy)-3,5-bis(trifluoromethyl)benzamide. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO (10mg/ml) or 100% ethanol (9mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0770