Learn More
TARGETMOL CHEMICALS INC CCG-1423 50MG
SDP
Catalog No. 502581041
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CCG-1423 a selective RhoA pathway inhibitor suppresses SRF-mediated transcription. purity: 99%
Catalog No. 50-258-1041 Supplier TARGETMOL CHEMICALS INC Supplier No. T201450MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CCG-1423 a selective RhoA pathway inhibitor suppresses SRF-mediated transcription. purity: 99%