Learn More
Abcam CCG 1423, Rho pathway-mediated signaling inhibitor, 25MG
Catalog No. 502672666
Encompass_Preferred
MW 454.7 Da, Purity >98%. Specific inhibitor of Rho pathway-mediated signaling and activation of serum response factor (SRF) transcription. Displays activity in several in vitro cancer cell functional assays. Potently (<1 mumol/L) inhibits lysophosphatidic acid-induced DNA synthesis in PC-3 prostate cancer cells, inhibits growth of RhoC-overexpressing melanoma lines at nanomolar concentrations. Selectively stimulates apoptosis of the metastasis-prone, RhoC-overexpressing melanoma cell lines. Acts at or downstream of MLK1 and upstream of SRF. Blocks MLK1 nuclear localization.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2666 Supplier Abcam Supplier No. AB22918125MG
MW 454.7 Da, Purity >98%. Specific inhibitor of Rho pathway-mediated signaling and activation of serum response factor (SRF) transcription. Displays activity in several in vitro cancer cell functional assays. Potently (<1 mumol/L) inhibits lysophosphatidic acid-induced DNA synthesis in PC-3 prostate cancer cells, inhibits growth of RhoC-overexpressing melanoma lines at nanomolar concentrations. Selectively stimulates apoptosis of the metastasis-prone, RhoC-overexpressing melanoma cell lines. Acts at or downstream of MLK1 and upstream of SRF. Blocks MLK1 nuclear localization.
The product is subject to the following: Abcam Restricted Use Statement