Learn More
Medchemexpress LLC HY-U00431 5mg Medchemexpress, CCG 203769 CAS:410074-60-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU004315MG
Encompass_Preferred
Medchemexpress, HY-U00431 5mg CCG 203769 CAS:410074-60-1 CCG 203769 is a selective G protein signaling ( RGS4 ) inhibitor, which blocks the RGS4-Gα o protein-protein interaction in vitro with an IC 50 of 17 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4444