Learn More
Medchemexpress LLC HY-108361 10mg Medchemexpress, CCG-203971 CAS:1443437-74-8 Purity:>98%
SDP
Catalog No. 501968540
Encompass_Preferred
Medchemexpress, HY-108361 10mg CCG-203971 CAS:1443437-74-8 CCG-203971 is a second-generation Rho/MRTF/SRF pathway inhibitor. CCG-203971 potently targets RhoA/C-activated SRE-luciferase (IC50 =6.4 μM). CCG-203971 inhibits PC-3 cell migration with an IC50 of 4.2 μM. Potential anti-metastasis Agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8540 Supplier Medchemexpress LLC Supplier No. HY10836110MG
Medchemexpress, HY-108361 10mg CCG-203971 CAS:1443437-74-8 CCG-203971 is a second-generation Rho/MRTF/SRF pathway inhibitor. CCG-203971 potently targets RhoA/C-activated SRE-luciferase (IC50 =6.4 μM). CCG-203971 inhibits PC-3 cell migration with an IC50 of 4.2 μM. Potential anti-metastasis Agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.