Learn More
Selleck Chemical LLC CCG-203971 10mg 1443437-74-8
SDP
Supplier:  Selleck Chemical LLC S846910MG
Encompass_Preferred
CCG-203971 is a novel small-molecule inhibitor of the Rho/MRTF/SRF pathway with the IC50 value of 0.64 ?M for SRE.L. It inhibits Rho-mediated gene transcription. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8204