Learn More
TARGETMOL CHEMICALS INC CCG-257081 5MG
Supplier: TARGETMOL CHEMICALS INC T307785MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CCG-257081 (MRTF/SRF-IN-1) is an inhibitor of cardiac myosin-related transcription factor and serum response factor (MRTF/SRF) that acts by inhibiting the Rho/MRTF/SRF pathway.CCG-257081 has shown efficacy in preventing bleomycin-induced fibrosis in several mouse models. CCG-257081 can be used in cancer prevention and fibrosis research. purity: 99%