Learn More
Medchemexpress LLC HY-13509 5mg , CCG-50014 CAS:883050-24-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135095MG
Encompass_Preferred
HY-13509 5mg CCG-50014 CAS: 883050-24-6 CCG-50014 is the most potent against the regulator of G-protein signaling protein type 4 (RGS4) (IC50 =30 nM) and is >20-fold selective for RGS4 over other RGS proteins. CCG-50014 binds covalently to the RGS, forming an adduct on two cysteine residues located in an allosteric regulatory site. CCG50014, reduces nociceptive responses and enhances opioid-mediated analgesic effects in the mouse formalin test. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1396