Learn More
Medchemexpress LLC HY-70074 5mg Medchemexpress, CCG-63802 CAS:620112-78-9 Purity:>98%
SDP
Catalog No. 502082272
Encompass_Preferred
Medchemexpress, HY-70074 5mg CCG-63802 CAS:620112-78-9 CCG-63802 is a selective, reversible and allosteric RGS4 inhibitor. CCG-63802 specifically binds to RGS4 and blocks the RGS4-Gαo interaction, with an IC50 value of 1.9 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2272 Supplier Medchemexpress LLC Supplier No. HY700745MG
Medchemexpress, HY-70074 5mg CCG-63802 CAS:620112-78-9 CCG-63802 is a selective, reversible and allosteric RGS4 inhibitor. CCG-63802 specifically binds to RGS4 and blocks the RGS4-Gαo interaction, with an IC50 value of 1.9 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.