Learn More
Bulldog Bio Inc CCHIP DISP HEMOCYTOMTR NI 20TS
SDP
Catalog No. 50999466
Encompass_Preferred
Consumable; Cell Counting;C-Chip Disposable Hemocytometer, Nuebuaer Improved Grid, 10 Slides, 2 tests/slide, 20 total tests.
Catalog No. 50-999-466 Supplier Bulldog Bio Inc Supplier No. DHCN002
Consumable; Cell Counting;C-Chip Disposable Hemocytometer, Nuebuaer Improved Grid, 10 Slides, 2 tests/slide, 20 total tests.