Learn More
Medchemexpress LLC CCI-006 10mg | 292053-42-0 | 10 MG
SDP
Catalog No. 5000308278
Encompass_Preferred
Methyl 2-cyano-3-(5-(4-sulfamoylphenyl)furan-2-yl)acrylate is a small-molecule mitochondrial respiration inhibitor that functions as a chemosensitizer in MLL-rearranged leukemia research; supplied as a light-yellow solid with high purity for preclinical laboratory use.
  • High purity suitable for research applications.
  • Molecular weight 332.33 g/mol; formula C15H12N2O5S.
  • Stable as powder at -20 °C for up to 3 years; in solvent store at -80 °C for up to 6 months.
  • Available in small pack sizes appropriate for in vitro studies.
  • Acts as a mitochondrial respiration inhibitor and chemosensitizer in MLL-r leukemia models.
Catalog No. 50-003-08278 Supplier Medchemexpress LLC Supplier No. HY11441010MG
Methyl 2-cyano-3-(5-(4-sulfamoylphenyl)furan-2-yl)acrylate is a small-molecule mitochondrial respiration inhibitor that functions as a chemosensitizer in MLL-rearranged leukemia research; supplied as a light-yellow solid with high purity for preclinical laboratory use.
  • High purity suitable for research applications.
  • Molecular weight 332.33 g/mol; formula C15H12N2O5S.
  • Stable as powder at -20 °C for up to 3 years; in solvent store at -80 °C for up to 6 months.
  • Available in small pack sizes appropriate for in vitro studies.
  • Acts as a mitochondrial respiration inhibitor and chemosensitizer in MLL-r leukemia models.