Learn More
Medchemexpress LLC CCI-006 50mg | 292053-42-0 | 50 MG
SDP
Catalog No. 5000227424
Encompass_Preferred
CCI-006 is a research-grade small molecule inhibitor of mitochondrial metabolism that selectively chemosensitizes a subset of MLL-rearranged leukemia cells by inducing mitochondrial depolarization and an unfolded protein response. Intended for laboratory research use only.
  • Inhibits mitochondrial respiration leading to mitochondrial depolarization.
  • Acts as a chemosensitizer in MLL-rearranged leukemia models.
  • High purity suitable for research applications.
  • Available in common research formats, including pre-made solutions for convenience.
  • Not for human or clinical use; for laboratory research only.
Catalog No. 50-002-27424 Supplier Medchemexpress LLC Supplier No. HY11441050MG
CCI-006 is a research-grade small molecule inhibitor of mitochondrial metabolism that selectively chemosensitizes a subset of MLL-rearranged leukemia cells by inducing mitochondrial depolarization and an unfolded protein response. Intended for laboratory research use only.
  • Inhibits mitochondrial respiration leading to mitochondrial depolarization.
  • Acts as a chemosensitizer in MLL-rearranged leukemia models.
  • High purity suitable for research applications.
  • Available in common research formats, including pre-made solutions for convenience.
  • Not for human or clinical use; for laboratory research only.