Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CCI-006 5mg | 292053-42-0 | 5 MG
SDP

Catalog No. 5000229940
Encompass_Preferred

CCI-006 is a research-grade small molecule that inhibits mitochondrial respiration and acts as a chemosensitizer in MLL-rearranged leukemia cells. Supplied as a high-purity solid for in vitro research use only.

  • High purity suitable for in vitro studies.
  • Molecular weight 332.33 g/mol.
  • Molecular formula C15H12N2O5S.
  • Light yellow to yellow solid physical form.
  • Intended for laboratory research use, not for human or diagnostic use.

Catalog No. 50-002-29940 Supplier Medchemexpress LLC Supplier No. HY1144105MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CCI-006 is a research-grade small molecule that inhibits mitochondrial respiration and acts as a chemosensitizer in MLL-rearranged leukemia cells. Supplied as a high-purity solid for in vitro research use only.

  • High purity suitable for in vitro studies.
  • Molecular weight 332.33 g/mol.
  • Molecular formula C15H12N2O5S.
  • Light yellow to yellow solid physical form.
  • Intended for laboratory research use, not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.