Learn More
Medchemexpress LLC Cci-007 | 939228-52-1 | 98.5% | 10 MG
SDP
Catalog No. 5000231301
Encompass_Preferred
CCI-007 is a research-grade small molecule reported to have cytotoxic activity against infant leukemia cells with MLL rearrangements, showing activity with reported IC50 values of 2.5-6.2 μM in sensitive cell lines. It is supplied at research purity for in vitro studies.
  • Reported IC50 2.5-6.2 μM in MLL-rearranged sensitive cells.
  • Manufacturer-stated purity approximately 98.5%.
  • Molecular formula C15H16N2O5S; molecular weight 336.36 g·mol⁻¹.
  • Available in small research quantities such as 10 MG for in vitro use.
  • For laboratory research use only; not for human or veterinary use.
Catalog No. 50-002-31301 Supplier Medchemexpress LLC Supplier No. HY12269810MG
CCI-007 is a research-grade small molecule reported to have cytotoxic activity against infant leukemia cells with MLL rearrangements, showing activity with reported IC50 values of 2.5-6.2 μM in sensitive cell lines. It is supplied at research purity for in vitro studies.
  • Reported IC50 2.5-6.2 μM in MLL-rearranged sensitive cells.
  • Manufacturer-stated purity approximately 98.5%.
  • Molecular formula C15H16N2O5S; molecular weight 336.36 g·mol⁻¹.
  • Available in small research quantities such as 10 MG for in vitro use.
  • For laboratory research use only; not for human or veterinary use.