Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CCI-007 | 939228-52-1 | 98.5% | 5 MG
SDP

Catalog No. 5000231302
Encompass_Preferred

CCI-007 is a small-molecule research reagent with cytotoxic activity against infant leukemia bearing MLL rearrangements. Reported IC50 values in sensitive cell lines are 2.5-6.2 μM, and the compound is supplied as a solid for laboratory research.

  • Cytotoxic activity against MLL-rearranged infant leukemia (IC50 2.5-6.2 μM).
  • Molecular formula C15H16N2O5S; molecular weight 336.36 g/mol.
  • Purity 98.45% as supplied.
  • Appearance: solid.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-31302 Supplier Medchemexpress LLC Supplier No. HY1226985MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CCI-007 is a small-molecule research reagent with cytotoxic activity against infant leukemia bearing MLL rearrangements. Reported IC50 values in sensitive cell lines are 2.5-6.2 μM, and the compound is supplied as a solid for laboratory research.

  • Cytotoxic activity against MLL-rearranged infant leukemia (IC50 2.5-6.2 μM).
  • Molecular formula C15H16N2O5S; molecular weight 336.36 g/mol.
  • Purity 98.45% as supplied.
  • Appearance: solid.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.