Learn More
Aviva Systems Biology Corp CCIN Antibody - N-terminal region (ARP53645_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53645_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCIN. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CCIN. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-862