Learn More
Aviva Systems Biology Corp CCIN Antibody - C-terminal region (ARP53646_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53646_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCIN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CCIN. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53723