Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CCK (26-31) sulfated | 89911-65-9 | 96.4% | 923.04 | 10 MG
SDP

Catalog No. 5000400283
Encompass_Preferred

CCK (26-31) sulfated is an N-terminal fragment of the peptide hormone CCK, which is found in the gut and brain. It plays a role in stimulating digestion, regulating satiety, and is associated with anxiety. This product is intended for laboratory research use.

  • N-terminal fragment of CCK
  • Peptide hormone found in the gut and brain
  • Stimulates digestion
  • Regulates satiety
  • Associated with anxiety
  • Appearance: White to off-white solid
  • Soluble in DMSO
  • Not classified as hazardous
  • Stable under recommended storage conditions

Catalog No. 50-004-00283 Supplier Medchemexpress LLC Supplier No. HYP267710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CCK (26-31) sulfated is an N-terminal fragment of the peptide hormone CCK, which is found in the gut and brain. It plays a role in stimulating digestion, regulating satiety, and is associated with anxiety. This product is intended for laboratory research use.

  • N-terminal fragment of CCK
  • Peptide hormone found in the gut and brain
  • Stimulates digestion
  • Regulates satiety
  • Associated with anxiety
  • Appearance: White to off-white solid
  • Soluble in DMSO
  • Not classified as hazardous
  • Stable under recommended storage conditions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.