Learn More
Medchemexpress LLC CCK (26-31) sulfated | 89911-65-9 | MFCD00133564 | 96.4% | 923.04 | 1 MG
SDP
Catalog No. 5000397345
Encompass_Preferred
CCK (26-31) sulfated is the N-terminal fragment of CCK, a peptide hormone found in the gut and brain. It stimulates digestion, regulates satiety, and is associated with anxiety. This product is for research use only.
  • N-terminal fragment of CCK
  • Stimulates digestion
  • Regulates satiety
  • Associated with anxiety
  • For research use only
Catalog No. 50-003-97345 Supplier Medchemexpress LLC Supplier No. HYP26771MG
CCK (26-31) sulfated is the N-terminal fragment of CCK, a peptide hormone found in the gut and brain. It stimulates digestion, regulates satiety, and is associated with anxiety. This product is for research use only.
  • N-terminal fragment of CCK
  • Stimulates digestion
  • Regulates satiety
  • Associated with anxiety
  • For research use only