Learn More
American Radiolabeled Chemicals Inc CCK-8-125I 10UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARI 013510UC
Encompass
Cholecystokinin octapeptide-125I (Bolton-Hunter labeled) 10uCi in buffered solution, 0.1 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~1415, Shipped in Dry Ice
Catalog No. 50-901-008
Add to Cart
Request a Quote