Learn More
American Radiolabeled Chemicals Inc CCK-8-125I 50UC
SDP
Catalog No. 50901009
Encompass
Cholecystokinin octapeptide-125I (Bolton-Hunter labeled) 50uCi in buffered solution, 0.1 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~1415, Shipped in Dry Ice
Catalog No. 50-901-009 Supplier American Radiolabeled Chemicals Inc Supplier No. ARI 013550UC
Add to Cart
Add to Cart
Request a Quote
Cholecystokinin octapeptide-125I (Bolton-Hunter labeled) 50uCi in buffered solution, 0.1 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~1415, Shipped in Dry Ice