Learn More
Apexbio Technology LLC Cell Counting Kit-8 (CCK-8), 2X5ml (1000 tests)
SDP
Catalog No. 501905563
Encompass_Preferred
Our product Cell Counting Kit-8 (CCK-8) provide a more convenient and sensitive method for the research of cell number determination and cell proliferationtotoxicity assay than previous ways. The kit utilizes a highly water-soluble tetrazolium salt, WST-8, which produces a water-soluble formazan dye upon reduction in the presence of an electron mediator. The amount of the formazan generated by dehydrogenases is directly in proportion to the numbers of living cells. The detection sensitivity by CCK-8 is higher than other tetrazolium salts such as MTT, XTT, MTS or WST-1. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5563 Supplier Apexbio Technology LLC Supplier No. K101810
Our product Cell Counting Kit-8 (CCK-8) provide a more convenient and sensitive method for the research of cell number determination and cell proliferationtotoxicity assay than previous ways. The kit utilizes a highly water-soluble tetrazolium salt, WST-8, which produces a water-soluble formazan dye upon reduction in the presence of an electron mediator. The amount of the formazan generated by dehydrogenases is directly in proportion to the numbers of living cells. The detection sensitivity by CCK-8 is higher than other tetrazolium salts such as MTT, XTT, MTS or WST-1. Other sizes are available. Please inqury us for quote.