Learn More
Aviva Systems Biology Corp CCK Antibody - middle region (ARP33849_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33849_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCK. Alternate names: MGC117187ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55171