Learn More
Eurofins Discoverx Ready-to-Assay CCK1 Cholesystokinin Receptor Frozen Cells, 2 Vials of Cells. (HTS184RTA)
Supplier:  Eurofins Discoverx HTS184RTA
Encompass
The Ready-to-Assay CCK1 Cholecystokinin Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. prostaglandin E2) based activation of CCKAR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CCKAR alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of CCKAR, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-6023