Learn More
Enzo Life Sciences Cholecystokinin-39 (9-20) polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences BMLCA11230100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to a portion of mammalian cholecystokinin-39. Source: Whole rabbit serum. Species reactivity: Human, Rat Porcine. Applications: IHC. Formulation: Liquid. Antiserum containing 0.1% sodium azide.
Catalog No. 50-201-0351