Learn More
Apexbio Technology LLC Cell Counting Kit-8 (CCK-8) Plus 1x1ml (100 tests)
SDP
Catalog No. 503107007
Encompass_Preferred
Cell Counting Kit-8 (CCK-8) Plus is a convenient and sensitive assay kit for cell proliferation and cytotoxicity detection It utilizes a highly water-soluble tetrazolium salt WST-8 which can be reduced by abundant dehydrogenases in living cells to a soluble orange-colored formazan dye in the presence of electron mediator reagents The amount of formazan produced by dehydrogenases is directly proportional to the number of viable cells This optimized kit requires only 0 5-1 hour incubation for detection offering greater sensitivity faster detection speed and a broader linear range compared with standard CCK-8 kits
Catalog No. 50-310-7007 Supplier Apexbio Technology LLC Supplier No. K22681
Cell Counting Kit-8 (CCK-8) Plus is a convenient and sensitive assay kit for cell proliferation and cytotoxicity detection It utilizes a highly water-soluble tetrazolium salt WST-8 which can be reduced by abundant dehydrogenases in living cells to a soluble orange-colored formazan dye in the presence of electron mediator reagents The amount of formazan produced by dehydrogenases is directly proportional to the number of viable cells This optimized kit requires only 0 5-1 hour incubation for detection offering greater sensitivity faster detection speed and a broader linear range compared with standard CCK-8 kits