Learn More
Aviva Systems Biology Corp CCKAR Antibody - N-terminal region (ARP64107_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64107_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCKAR. It was validated on Western Blot. . Alternate names: CCK-A, CCK1-R, CCKRAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-177