Learn More
Enzo Life Sciences CCL1 (human) monoclonal antibody (8P16-N4-L21) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804489C100
Encompass_Preferred
Clone: 8P16-N4-L21. Isotype: IgM. Immunogen: Recombinant human CCL1 (I-309). Species reactivity: Human. Applications: IHC (FS). Purity: DEAE purified. Formulation: Liquid. In PBS.
Catalog No. 50-200-9922