Learn More
Aviva Systems Biology Corp CCL13 Antibody - middle region (AVARP07007_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP07007_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCL13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCL13. Alternate names: CKb10, MCP-4, MGC17134, NCC-1, NCC1, SCYA13, SCYL1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62543