Learn More
ABclonal Technology CCL17 Rabbit mAb
SDP
Catalog No. 5000010302
Encompass_Preferred
This antimicrobial gene is one of several Cys-Cys (CC) cytokine genes clustered on the q arm of chromosome 16 Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes The CC cytokines are proteins characterized by two adjacent cysteines The cytokine encoded by this gene displays chemotactic activity for T lymphocytes but not monocytes or granulocytes The product of this gene binds to chemokine receptors CCR4 and CCR8 This chemokine plays important roles in T cell development in thymus as well as in trafficking and activation of mature T cells
Catalog No. 50-000-10302 Supplier ABclonal Technology Supplier No. A25954100UL
This antimicrobial gene is one of several Cys-Cys (CC) cytokine genes clustered on the q arm of chromosome 16 Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes The CC cytokines are proteins characterized by two adjacent cysteines The cytokine encoded by this gene displays chemotactic activity for T lymphocytes but not monocytes or granulocytes The product of this gene binds to chemokine receptors CCR4 and CCR8 This chemokine plays important roles in T cell development in thymus as well as in trafficking and activation of mature T cells